Product Details

SNP ID
rs138413849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3811975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGCCACAACGTGGGGACAGAAA[G/T]AATAAATATCAATGTATAACTTCTA
Phenotype
MIM: 604682 MIM: 616624
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGAE PubMed Links

Gene Details

Gene
ITGAE
Gene Name
integrin subunit alpha E
There are no transcripts associated with this gene.

Gene
NCBP3
Gene Name
nuclear cap binding subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114118.2 2415 UTR 3 NP_001107590.1
XM_005256717.2 2415 UTR 3 XP_005256774.1
XM_006721556.2 2415 UTR 3 XP_006721619.1
XM_011523954.2 2415 UTR 3 XP_011522256.1
XM_011523955.2 2415 UTR 3 XP_011522257.1
XM_017024826.1 2415 UTR 3 XP_016880315.1
XM_017024827.1 2415 UTR 3 XP_016880316.1
XM_017024828.1 2415 UTR 3 XP_016880317.1
XM_017024829.1 2415 UTR 3 XP_016880318.1

View Full Product Details