Product Details

SNP ID
rs138644325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29562776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGCTCCAGGCTGGCAAGGGCT[C/G]CTGGCGCAGGAAGCCACAGTGGCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198147.2 1250 Missense Mutation CAG,GAG Q398E NP_937790.2
Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
There are no transcripts associated with this gene.

View Full Product Details