Product Details

SNP ID
rs139053410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9901006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCACCATCATCATTCTTTCCA[A/C]AGTACTTCCAGATCTTCTCGGCTCG
Phenotype
MIM: 603127 MIM: 603659 MIM: 179618
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GAS7 PubMed Links

Gene Details

Gene
GAS7
Gene Name
growth arrest specific 7
There are no transcripts associated with this gene.

Gene
GLP2R
Gene Name
glucagon like peptide 2 receptor
There are no transcripts associated with this gene.

Gene
RCVRN
Gene Name
recoverin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002903.2 663 Missense Mutation TGT,TTT C159F NP_002894.1

View Full Product Details