Product Details

SNP ID
rs139142062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41825638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGATGTCATAGGAGTCCATGTAG[C/T]GCTCCCGCCGCTCCTCCACCTGCCC
Phenotype
MIM: 607063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
There are no transcripts associated with this gene.

Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052935.4 866 Missense Mutation CAC,CGC H263R NP_443167.4
XM_006721669.3 866 Missense Mutation CAC,CGC H220R XP_006721732.1
XM_006721670.3 866 Missense Mutation CAC,CGC H220R XP_006721733.1
XM_011524276.2 866 Missense Mutation CAC,CGC H196R XP_011522578.1
XM_011524277.2 866 Missense Mutation CAC,CGC H188R XP_011522579.1
XM_017024127.1 866 UTR 3 XP_016879616.1

View Full Product Details