Product Details

SNP ID
rs139380300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4741012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCCCCTACCAGCAGGGAGTCCC[C/T]CCAGGAAACAAACCCTCCAGGAGAG
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
There are no transcripts associated with this gene.

Gene
MED11
Gene Name
mediator complex subunit 11
There are no transcripts associated with this gene.

Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136046.2 530 Missense Mutation CCC,CTC P155L NP_001129518.1
NM_001267822.1 530 Missense Mutation CCC,CTC P155L NP_001254751.1
NM_032265.2 530 Missense Mutation CCC,CTC P155L NP_115641.1
XM_017025218.1 530 Missense Mutation CCC,CTC P155L XP_016880707.1

View Full Product Details