Product Details

SNP ID
rs139574677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7313062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGCATCTGCACAGGGAGCTG[C/G]GGGGAAGCAAGGAGTCCAGGGGCTG
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143760.1 943 Intron NP_001137232.1
NM_001143761.1 943 Intron NP_001137233.1
NM_001143762.1 943 Intron NP_001137234.1
NM_001970.4 943 Intron NP_001961.1
XM_005256509.2 943 Intron XP_005256566.1
XM_011523710.2 943 Intron XP_011522012.1
XM_011523711.2 943 Intron XP_011522013.1
XM_011523712.2 943 Intron XP_011522014.1
XM_011523713.2 943 Intron XP_011522015.1
XM_017024300.1 943 Intron XP_016879789.1
XM_017024301.1 943 Intron XP_016879790.1
Gene
GPS2
Gene Name
G protein pathway suppressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004489.4 943 Silent Mutation CCC,CCG P289P NP_004480.1
Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details