Product Details

SNP ID
rs139687237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63606376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGTGGCTCTTCTCTGCTCATC[A/G]AGGCATTATCTAACAGTAGCCACAA
Phenotype
MIM: 612958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TACO1 PubMed Links

Gene Details

Gene
TACO1
Gene Name
translational activator of cytochrome c oxidase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016360.3 663 Missense Mutation AAG,GAG K151E NP_057444.2

View Full Product Details