Product Details

SNP ID
rs139806967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76735973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTACGAAACAGCCATTATTAT[C/T]TAAGCTTCATACACATCCTTTCTGC
Phenotype
MIM: 604914 MIM: 615262 MIM: 600813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD6 PubMed Links

Gene Details

Gene
JMJD6
Gene Name
arginine demethylase and lysine hydroxylase
There are no transcripts associated with this gene.

Gene
METTL23
Gene Name
methyltransferase like 23
There are no transcripts associated with this gene.

Gene
MFSD11
Gene Name
major facilitator superfamily domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242532.1 1105 Intron NP_001229461.1
NM_001242533.1 1105 Intron NP_001229462.1
NM_001242534.1 1105 Intron NP_001229463.1
NM_001242535.1 1105 Intron NP_001229464.1
NM_001242536.1 1105 Intron NP_001229465.1
NM_001242537.1 1105 Intron NP_001229466.1
NM_024311.3 1105 Intron NP_077287.1
XM_011525236.2 1105 Intron XP_011523538.1
XM_011525237.2 1105 Intron XP_011523539.2
XM_011525238.2 1105 Intron XP_011523540.2
XM_011525239.2 1105 Intron XP_011523541.2
XM_011525240.2 1105 Intron XP_011523542.2
XM_011525241.2 1105 Intron XP_011523543.2
XM_011525242.1 1105 Intron XP_011523544.1
XM_011525244.1 1105 Intron XP_011523546.1
XM_011525247.2 1105 Intron XP_011523549.2
XM_017025065.1 1105 Intron XP_016880554.1
XM_017025066.1 1105 Intron XP_016880555.1
XM_017025067.1 1105 Intron XP_016880556.1
XM_017025068.1 1105 Intron XP_016880557.1
XM_017025069.1 1105 Intron XP_016880558.1
XM_017025070.1 1105 Intron XP_016880559.1
XM_017025071.1 1105 Intron XP_016880560.1
Gene
MIR636
Gene Name
microRNA 636
There are no transcripts associated with this gene.

Gene
SRSF2
Gene Name
serine and arginine rich splicing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195427.1 1105 Intron NP_001182356.1
NM_003016.4 1105 UTR 3 NP_003007.2
XM_017024942.1 1105 Intron XP_016880431.1

View Full Product Details