Product Details

SNP ID
rs139957500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7855508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGACCCCGGTTTCACGGCCCTA[C/T]TGGTCACCGGATTCCTGCTCCTCGT
Phenotype
MIM: 611577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5D1 PubMed Links

Gene Details

Gene
CYB5D1
Gene Name
cytochrome b5 domain containing 1
There are no transcripts associated with this gene.

Gene
KDM6B
Gene Name
lysine demethylase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080424.1 283 Intron NP_001073893.1
XM_005256549.3 283 Intron XP_005256606.1
XM_005256550.4 283 Intron XP_005256607.1
XM_005256551.3 283 Intron XP_005256608.1
XM_005256552.4 283 Intron XP_005256609.1
XM_006721483.3 283 Intron XP_006721546.1
XM_011523750.2 283 Intron XP_011522052.1
XM_011523752.2 283 Intron XP_011522054.1
XM_017024381.1 283 Intron XP_016879870.1
Gene
NAA38
Gene Name
N(alpha)-acetyltransferase 38, NatC auxiliary subunit
There are no transcripts associated with this gene.

Gene
TMEM88
Gene Name
transmembrane protein 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319941.1 283 Intron NP_001306870.1
NM_203411.1 283 Silent Mutation CTG,TTG L92L NP_981956.1
XM_005256856.3 283 Silent Mutation CTG,TTG L106L XP_005256913.1

View Full Product Details