Product Details

SNP ID
rs140033860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42574156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCACCTTAGCAGTGAGGGCCA[C/T]GATTTTGCCATCTAGGACTTGAAGG
Phenotype
MIM: 609855 MIM: 616498 MIM: 602976 MIM: 608665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COASY PubMed Links

Gene Details

Gene
COASY
Gene Name
Coenzyme A synthase
There are no transcripts associated with this gene.

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
There are no transcripts associated with this gene.

Gene
MLX
Gene Name
MLX, MAX dimerization protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170607.2 425 Intron NP_733752.1
NM_198204.1 425 Intron NP_937847.1
NM_198205.1 425 Intron NP_937848.1
XM_017024991.1 425 Intron XP_016880480.1
Gene
PSMC3IP
Gene Name
PSMC3 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256014.1 425 Missense Mutation ATG,GTG M31V NP_001242943.1
NM_001256015.1 425 Missense Mutation ATG,GTG M15V NP_001242944.1
NM_001256016.1 425 Missense Mutation ATG,GTG M15V NP_001242945.1
NM_013290.6 425 Missense Mutation ATG,GTG M94V NP_037422.2
NM_016556.3 425 Missense Mutation ATG,GTG M94V NP_057640.1

View Full Product Details