Product Details
- SNP ID
-
rs140312984
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8289580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACTGATCTCTTGCTTACCTTCAA[C/T]CAGCCCCCGTAAGGAGGAAGGAACG
- Phenotype
-
MIM: 607954
MIM: 610818
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RANGRF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs869773] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RANGRF
- Gene Name
- RAN guanine nucleotide release factor
- Gene
- SLC25A35
- Gene Name
- solute carrier family 25 member 35
View Full Product Details