Product Details

SNP ID
rs140415109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37033808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAGATGGCTGTTGCAACTTAA[C/T]GCCATCAAACATGCATTCAAGGCAG
Phenotype
MIM: 608463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AATF PubMed Links

Gene Details

Gene
AATF
Gene Name
apoptosis antagonizing transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012138.3 Intron NP_036270.1
XM_011524611.1 Intron XP_011522913.1
Gene
MIR2909
Gene Name
microRNA 2909
There are no transcripts associated with this gene.

View Full Product Details