Product Details

SNP ID
rs140504068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1271262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCGCCCCCGGGATGGGCCATCC[A/G]CGCTCCTGACCGGCCTCGAGGCACC
Phenotype
MIM: 615416 MIM: 612211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHLHA9 PubMed Links

Gene Details

Gene
BHLHA9
Gene Name
basic helix-loop-helix family member a9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164405.1 699 Silent Mutation CCA,CCG P233P NP_001157877.1
Gene
TUSC5
Gene Name
tumor suppressor candidate 5
There are no transcripts associated with this gene.

View Full Product Details