Product Details

SNP ID
rs140740946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40863197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCGGGCCTTGACATTCAGCAG[C/T]CGCTGGTGGTCCACGTTCTGGCGCT
Phenotype
MIM: 601687
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT12 PubMed Links

Gene Details

Gene
KRT12
Gene Name
keratin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000223.3 1266 Silent Mutation CGA,CGG R414R NP_000214.1

View Full Product Details