Product Details

SNP ID
rs140926091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3923935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGTCAGCAGGTGGGTACAGGG[C/T]CCGCTCCTGCTAAGACCTCCCCACC
Phenotype
MIM: 601929 MIM: 600845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2A3 PubMed Links

Gene Details

Gene
ATP2A3
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005173.3 4806 UTR 3 NP_005164.2
NM_174953.2 4806 UTR 3 NP_777613.1
NM_174954.2 4806 UTR 3 NP_777614.1
NM_174955.2 4806 UTR 3 NP_777615.1
NM_174956.2 4806 UTR 3 NP_777616.1
NM_174957.2 4806 UTR 3 NP_777617.1
NM_174958.2 4806 UTR 3 NP_777618.1
XM_011523881.2 4806 Intron XP_011522183.1
XM_011523882.2 4806 Intron XP_011522184.1
XM_011523884.2 4806 Intron XP_011522186.1
XM_011523885.1 4806 UTR 3 XP_011522187.1
XM_011523888.2 4806 UTR 3 XP_011522190.1
XM_011523889.1 4806 Intron XP_011522191.1
XM_011523892.2 4806 Intron XP_011522194.1
XM_017024692.1 4806 Intron XP_016880181.1
XM_017024693.1 4806 Intron XP_016880182.1
Gene
P2RX1
Gene Name
purinergic receptor P2X 1
There are no transcripts associated with this gene.

View Full Product Details