Product Details

SNP ID
rs141035572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTTGTCCTTGGGCTCTGGGCC[A/G]TCTTCTGTAACAACTGACAGAAAAT
Phenotype
MIM: 604334 MIM: 616463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 784 Silent Mutation GAC,GAT D213D NP_001307881.1
NM_001320953.1 784 Silent Mutation GAC,GAT D195D NP_001307882.1
NM_001320954.1 784 Silent Mutation GAC,GAT D195D NP_001307883.1
NM_001320955.1 784 Silent Mutation GAC,GAT D194D NP_001307884.1
NM_014519.3 784 Silent Mutation GAC,GAT D222D NP_055334.2
XM_011524006.2 784 Silent Mutation GAC,GAT D174D XP_011522308.1
XM_017025021.1 784 Silent Mutation GAC,GAT D222D XP_016880510.1
XM_017025022.1 784 Silent Mutation GAC,GAT D146D XP_016880511.1
XM_017025023.1 784 Silent Mutation GAC,GAT D137D XP_016880512.1

View Full Product Details