Product Details

SNP ID
rs141260911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69150008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCCTCTAACTTGAAAAAGGCCC[A/G]AGATAGAGGGTGGACATCCTCCACA
Phenotype
MIM: 612508 MIM: 612504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080282.3 5332 Missense Mutation CGG,TGG R1485W NP_525021.3
Gene
ABCA6
Gene Name
ATP binding cassette subfamily A member 6
There are no transcripts associated with this gene.

View Full Product Details