Product Details

SNP ID
rs141685218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34361871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTCTTGCTCCGCAAATGAGAA[A/G]CAACATCTGGAGAAGGGTACCTGCA
Phenotype
MIM: 182281 MIM: 601391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL1 PubMed Links

Gene Details

Gene
CCL1
Gene Name
C-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002981.2 174 Silent Mutation TGC,TGT C34C NP_002972.1
Gene
CCL13
Gene Name
C-C motif chemokine ligand 13
There are no transcripts associated with this gene.

View Full Product Details