Product Details

SNP ID
rs141797289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69418439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTAAAAGAAATTTCATAAATAA[C/T]CCTTGTTCATTATAGAAACATTTCA
Phenotype
MIM: 601254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP2K6 PubMed Links

Gene Details

Gene
MAP2K6
Gene Name
mitogen-activated protein kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002758.3 Intron NP_002749.2
XM_005257515.2 Intron XP_005257572.1
XM_005257516.2 Intron XP_005257573.1
XM_006721975.3 Intron XP_006722038.1
XM_011525025.2 Intron XP_011523327.1
XM_011525026.2 Intron XP_011523328.1
XM_011525027.2 Intron XP_011523329.1

View Full Product Details