Product Details

SNP ID
rs141992383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62705543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGGCCCCCAAAACCTACTTTC[A/G]TTTTCCTCTGTTCTGGGTTGTGGGT
Phenotype
MIM: 613337
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371855 PubMed Links

Gene Details

Gene
LOC105371855
Gene Name
uncharacterized LOC105371855
There are no transcripts associated with this gene.

Gene
MARCH10
Gene Name
membrane associated ring-CH-type finger 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100875.2 2803 Silent Mutation AAC,AAT N789N NP_001094345.1
NM_001288779.1 2803 Silent Mutation AAC,AAT N827N NP_001275708.1
NM_001288780.1 2803 Silent Mutation AAC,AAT N788N NP_001275709.1
NM_152598.3 2803 Silent Mutation AAC,AAT N789N NP_689811.2
XM_005257095.2 2803 Intron XP_005257152.2
XM_005257096.2 2803 Intron XP_005257153.2
XM_005257097.3 2803 Intron XP_005257154.2
XM_005257098.2 2803 Intron XP_005257155.2
XM_005257100.2 2803 Intron XP_005257157.2
XM_005257102.2 2803 Intron XP_005257159.2
XM_005257103.1 2803 Silent Mutation AAC,AAT N788N XP_005257160.1
XM_005257106.4 2803 Intron XP_005257163.2
XM_011524435.1 2803 Intron XP_011522737.1
XM_011524436.1 2803 Intron XP_011522738.1
XM_011524437.1 2803 Silent Mutation AAC,AAT N788N XP_011522739.1

View Full Product Details