Product Details

SNP ID
rs142272467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35409491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGCCAGATTGCAAGGACTCCT[C/T]GTATATCCACTTGTGTTCTTCCTTC
Phenotype
MIM: 614955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLFN12 PubMed Links

Gene Details

Gene
SLFN12
Gene Name
schlafen family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289009.1 3852 Intron NP_001275938.1
NM_018042.4 3852 Intron NP_060512.3
XM_005257995.4 3852 UTR 3 XP_005258052.1
XM_011524967.2 3852 UTR 3 XP_011523269.1
XM_017024809.1 3852 UTR 3 XP_016880298.1
XM_017024810.1 3852 UTR 3 XP_016880299.1
XM_017024811.1 3852 Intron XP_016880300.1

View Full Product Details