Product Details

SNP ID
rs142323373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34320333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATAGGAAAATTCCTATCCAGAG[A/G]CTGGAGAGCTACACAAGAATCACCA
Phenotype
MIM: 602283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL8 PubMed Links

Gene Details

Gene
CCL8
Gene Name
C-C motif chemokine ligand 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005623.2 596 Silent Mutation AGA,AGG R47R NP_005614.2

View Full Product Details