Product Details

SNP ID
rs142348643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21238909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATCTCTTTAATATCTTGTAGC[C/T]GTAAGACTGATACAACTGAAAACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NATD1 PubMed Links

Gene Details

Gene
NATD1
Gene Name
N-acetyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152914.2 4748 UTR 3 NP_690878.2

View Full Product Details