Product Details

SNP ID
rs142726996
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35576069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTTCTTGATTTTCAGATGAGT[A/G]CCACCAGTCAAGGAACTGCAAGAAG
Phenotype
MIM: 601758
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC105371743 PubMed Links

Gene Details

Gene
LOC105371743
Gene Name
uncharacterized LOC105371743
There are no transcripts associated with this gene.

Gene
PEX12
Gene Name
peroxisomal biogenesis factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000286.2 1409 Missense Mutation CAC,TAC H265Y NP_000277.1
Gene
SNORD7
Gene Name
small nucleolar RNA, C/D box 7
There are no transcripts associated with this gene.

View Full Product Details