Product Details

SNP ID
rs142786789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38365375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGGGGTCTTCCTTGCAGTCTTT[C/T]CCCCTACCTCCGCCTCCTCCACCCC
Phenotype
MIM: 608788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOCS7 PubMed Links

Gene Details

Gene
SOCS7
Gene Name
suppressor of cytokine signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014598.3 1147 Silent Mutation TTC,TTT F342F NP_055413.1
XM_017024551.1 1147 Intron XP_016880040.1
XM_017024552.1 1147 Intron XP_016880041.1

View Full Product Details