Product Details

SNP ID
rs142930242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39944069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCTCCCCGGGGCTGTTATGTG[G/T]CAAAGGAAGCAGGTGAACGGACGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC3C PubMed Links

Gene Details

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195545.1 378 Missense Mutation GCA,TCA A55S NP_001182474.1
XM_011524158.2 378 Missense Mutation GCA,TCA A55S XP_011522460.1
XM_017024003.1 378 Missense Mutation GCA,TCA A55S XP_016879492.1

View Full Product Details