Product Details

SNP ID
rs143092701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6425609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGTGGCTCTGTGGGTGGCTCTG[C/T]GGGAGGCTGCGTGGCACCCTGGCTC
Phenotype
MIM: 604392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIPL1 PubMed Links

Gene Details

Gene
AIPL1
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033054.2 1029 Missense Mutation ACA,GCA T273A NP_001028226.1
NM_001033055.2 1029 Missense Mutation ACA,GCA T276A NP_001028227.1
NM_001285399.2 1029 Missense Mutation ACA,GCA T324A NP_001272328.1
NM_001285400.2 1029 Missense Mutation ACA,GCA T314A NP_001272329.1
NM_001285401.2 1029 Missense Mutation ACA,GCA T312A NP_001272330.1
NM_001285402.1 1029 Missense Mutation ACA,GCA T297A NP_001272331.1
NM_001285403.2 1029 Intron NP_001272332.1
NM_014336.4 1029 Missense Mutation ACA,GCA T336A NP_055151.3

View Full Product Details