Product Details

SNP ID
rs143106589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19385234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCAAAGTCCTCCAAGTCCACTC[A/G]CAGCTCATACTTCTGCTTCAGTGTC
Phenotype
MIM: 614144 MIM: 602521 MIM: 600596
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B9D1 PubMed Links

Gene Details

Gene
B9D1
Gene Name
B9 domain containing 1
There are no transcripts associated with this gene.

Gene
MAPK7
Gene Name
mitogen-activated protein kinase 7
There are no transcripts associated with this gene.

Gene
MFAP4
Gene Name
microfibrillar associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198695.1 449 Nonsense Mutation CGA,TGA R153* NP_001185624.1
NM_002404.2 449 Nonsense Mutation CGA,TGA R129* NP_002395.1

View Full Product Details