Product Details

SNP ID
rs143353275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28547308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGCCGTGGGGTCAGGGTGTCC[C/T]GCTGTAGGAATAGTCTGCTTTATTG
Phenotype
MIM: 600838 MIM: 610271 MIM: 604011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXN1 PubMed Links

Gene Details

Gene
FOXN1
Gene Name
forkhead box N1
There are no transcripts associated with this gene.

Gene
PIGS
Gene Name
phosphatidylinositol glycan anchor biosynthesis class S
There are no transcripts associated with this gene.

Gene
UNC119
Gene Name
unc-119 lipid binding chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005148.3 1050 Missense Mutation NP_005139.1
NM_054035.2 1050 UTR 3 NP_473376.1
XM_011525459.2 1050 Intron XP_011523761.1
XM_017025289.1 1050 Missense Mutation XP_016880778.1

View Full Product Details