Product Details

SNP ID
rs143433100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50535164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGGGGTACCAGAGGTCCACT[A/G]TGTGTCCAGCGTTTTGGGGCTTTGG
Phenotype
MIM: 607264 MIM: 609835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPN3 PubMed Links

Gene Details

Gene
EPN3
Gene Name
epsin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017957.2 Intron NP_060427.2
Gene
LOC105371824
Gene Name
uncharacterized LOC105371824
There are no transcripts associated with this gene.

Gene
MYCBPAP
Gene Name
MYCBP associated protein
There are no transcripts associated with this gene.

View Full Product Details