Product Details
- SNP ID
-
rs143659814
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81510789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGGACAGTGAGGCCAGGATGGA[A/G]CCACCGATCCACACCGAGTACTTGC
- Phenotype
-
MIM: 102560
MIM: 607643
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1139406] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTG1
- Gene Name
- actin gamma 1
- Gene
- FSCN2
- Gene Name
- fascin actin-bundling protein 2, retinal
There are no transcripts associated with this gene.
View Full Product Details