Product Details

SNP ID
rs143745130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82049046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACTCACCGTGTGGGGTCTTGA[G/T]GCAGGCTGAAGCCTCCAGCCACGTT
Phenotype
MIM: 601934 MIM: 602578
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DUS1L PubMed Links

Gene Details

Gene
DUS1L
Gene Name
dihydrouridine synthase 1 like
There are no transcripts associated with this gene.

Gene
GPS1
Gene Name
G protein pathway suppressor 1
There are no transcripts associated with this gene.

Gene
RFNG
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002917.1 907 Missense Mutation CAT,CCT H300P NP_002908.1
XM_011523587.2 907 Missense Mutation CAT,CCT H174P XP_011521889.1

View Full Product Details