Product Details
- SNP ID
-
rs143785213
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81191031 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACCGCTCCTGAAGCTTCCGCTCC[A/G]ACTGCTCCAGCTCGGAGAGCTCGGC
- Phenotype
-
MIM: 613479
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AATK-AS1
PubMed Links
Gene Details
- Gene
- AATK-AS1
- Gene Name
- AATK antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- CEP131
- Gene Name
- centrosomal protein 131
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009811.3 |
3072 |
Missense Mutation |
TCG,TTG |
S904L |
NP_001009811.2 |
NM_001319228.1 |
3072 |
Missense Mutation |
TCG,TTG |
S943L |
NP_001306157.1 |
NM_001319229.1 |
3072 |
Missense Mutation |
TCG,TTG |
S907L |
NP_001306158.1 |
NM_014984.3 |
3072 |
Missense Mutation |
TCG,TTG |
S940L |
NP_055799.2 |
XM_011524547.2 |
3072 |
Missense Mutation |
TCG,TTG |
S948L |
XP_011522849.1 |
XM_011524548.2 |
3072 |
Missense Mutation |
TCG,TTG |
S945L |
XP_011522850.1 |
XM_011524550.2 |
3072 |
Missense Mutation |
TCG,TTG |
S942L |
XP_011522852.1 |
XM_011524551.2 |
3072 |
Missense Mutation |
TCG,TTG |
S948L |
XP_011522853.1 |
XM_011524552.2 |
3072 |
Missense Mutation |
TCG,TTG |
S912L |
XP_011522854.1 |
XM_011524553.2 |
3072 |
Missense Mutation |
TCG,TTG |
S907L |
XP_011522855.1 |
XM_011524555.2 |
3072 |
Missense Mutation |
TCG,TTG |
S948L |
XP_011522857.1 |
XM_017024369.1 |
3072 |
Missense Mutation |
TCG,TTG |
S943L |
XP_016879858.1 |
View Full Product Details