Product Details

SNP ID
rs143878413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58491741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACGAGAGACTTGAATGTGTTC[A/G]TAACATGAGTTACAGACGAGAACTG
Phenotype
MIM: 603559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSF5 PubMed Links

Gene Details

Gene
HSF5
Gene Name
heat shock transcription factor 5
There are no transcripts associated with this gene.

Gene
MTMR4
Gene Name
myotubularin related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004687.4 3737 Silent Mutation TAC,TAT Y1170Y NP_004678.3
XM_005257784.2 3737 Silent Mutation TAC,TAT Y1184Y XP_005257841.1
XM_005257785.4 3737 Silent Mutation TAC,TAT Y1174Y XP_005257842.1
XM_005257786.4 3737 Silent Mutation TAC,TAT Y1170Y XP_005257843.1
XM_006722168.3 3737 Silent Mutation TAC,TAT Y1170Y XP_006722231.1
XM_011525460.2 3737 Silent Mutation TAC,TAT Y1174Y XP_011523762.1

View Full Product Details