Product Details

SNP ID
rs144036152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7220040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGCAGCTGCTGAGGCTCGGGG[A/G]CGGAAGGTCTGTGTGTGACAAGAGG
Phenotype
MIM: 609575 MIM: 602887 MIM: 602151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACADVL PubMed Links

Gene Details

Gene
ACADVL
Gene Name
acyl-CoA dehydrogenase, very long chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000018.3 11 Missense Mutation GAC,GGC D19G NP_000009.1
NM_001033859.2 11 Missense Mutation GAC,GGC D19G NP_001029031.1
NM_001270447.1 11 Intron NP_001257376.1
NM_001270448.1 11 UTR 5 NP_001257377.1
XM_006721516.2 11 Missense Mutation GAC,GGC D19G XP_006721579.2
XM_011523829.1 11 Missense Mutation GAC,GGC D19G XP_011522131.1
XM_011523830.1 11 Missense Mutation GAC,GGC D19G XP_011522132.1
Gene
DLG4
Gene Name
discs large MAGUK scaffold protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128827.2 11 Intron NP_001122299.1
NM_001321074.1 11 UTR 5 NP_001308003.1
NM_001321075.1 11 Intron NP_001308004.1
NM_001321076.1 11 Intron NP_001308005.1
NM_001321077.1 11 Intron NP_001308006.1
NM_001365.4 11 UTR 5 NP_001356.1
XM_005256491.1 11 Intron XP_005256548.1
XM_011523698.1 11 Intron XP_011522000.1
XM_011523699.2 11 Intron XP_011522001.1
XM_011523702.1 11 Intron XP_011522004.1
XM_017024288.1 11 Intron XP_016879777.1
XM_017024289.1 11 Intron XP_016879778.1
XM_017024290.1 11 Intron XP_016879779.1
Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
There are no transcripts associated with this gene.

Gene
MIR324
Gene Name
microRNA 324
There are no transcripts associated with this gene.

View Full Product Details