Product Details

SNP ID
rs144114953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81510732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGATGGAGGGGCCCGACTCGTC[A/G]TACTCCTGCTTGCTAATCCACATCT
Phenotype
MIM: 102560 MIM: 607643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTG1 PubMed Links

Gene Details

Gene
ACTG1
Gene Name
actin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199954.1 1225 Silent Mutation TAC,TAT Y362Y NP_001186883.1
NM_001614.3 1225 Silent Mutation TAC,TAT Y362Y NP_001605.1
Gene
FSCN2
Gene Name
fascin actin-bundling protein 2, retinal
There are no transcripts associated with this gene.

View Full Product Details