Product Details
- SNP ID
-
rs144276670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:79795156 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCTCCGAGGCCGGCGCTGGGTT[C/T]GCCTAAGGGCCTCTGTGAGGGGTCC
- Phenotype
-
MIM: 602770
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBX2
PubMed Links
Gene Details
- Gene
- CBX2
- Gene Name
- chromobox 2
There are no transcripts associated with this gene.
- Gene
- CBX8
- Gene Name
- chromobox 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020649.2 |
767 |
Missense Mutation |
AAA,GAA |
K217E |
NP_065700.1 |
View Full Product Details