Product Details

SNP ID
rs144404351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43781846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCAGAAAATCGCAAATGCCCAT[C/T]GCGGCCCTGCCAAGGTGGATTGCTG
Phenotype
MIM: 600183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf105 PubMed Links

Gene Details

Gene
C17orf105
Gene Name
chromosome 17 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136483.1 296 Missense Mutation CGC,TGC R90C NP_001129955.1
XM_011524633.1 296 Missense Mutation CGC,TGC R90C XP_011522935.1
XM_011524634.2 296 Missense Mutation CGC,TGC R90C XP_011522936.1
XM_011524635.1 296 Missense Mutation CGC,TGC R90C XP_011522937.1
XM_011524636.1 296 Missense Mutation CGC,TGC R90C XP_011522938.1
Gene
DUSP3
Gene Name
dual specificity phosphatase 3
There are no transcripts associated with this gene.

View Full Product Details