Product Details

SNP ID
rs144541706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28331431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATTTCCAGCCAAAAGATGACTT[C/T]AGTAAGTCTGTTGAGCTGCTTCTAG
Phenotype
MIM: 614394 MIM: 612912 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267774.1 Intron NP_001254703.1
NM_001267775.1 Intron NP_001254704.1
NM_001267776.1 Intron NP_001254705.1
NM_001267777.1 Intron NP_001254706.1
NM_001267778.1 Intron NP_001254707.1
NM_174887.3 Intron NP_777547.1
Gene
TMEM97
Gene Name
transmembrane protein 97
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details