Product Details

SNP ID
rs144718253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67044688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAGACCAAAATGCTGAAGGTC[C/T]GCGTGACCCTCTTCTGCATCCTGGC
Phenotype
MIM: 114209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNG1 PubMed Links

Gene Details

Gene
CACNG1
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000727.3 153 Missense Mutation CGC,TGC R10C NP_000718.1

View Full Product Details