Product Details

SNP ID
rs144837294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44657220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGACGTCTTCGGCAGCGTGATG[C/T]TGACTGGCTCCGCTTCCTTCTACGA
Phenotype
MIM: 616934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01180 PubMed Links

Gene Details

Gene
LINC01180
Gene Name
long intergenic non-protein coding RNA 1180
There are no transcripts associated with this gene.

Gene
MEIOC
Gene Name
meiosis specific with coiled-coil domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145080.2 360 Silent Mutation CTG,TTG L55L NP_001138552.2
XM_005257236.3 360 Silent Mutation CTG,TTG L55L XP_005257293.1

View Full Product Details