Product Details

SNP ID
rs144858776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75110306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGGCCATGGCCCAGAAGCCGAA[A/G]GTGGACCCCCACGTCGGGCGGCTGG
Phenotype
MIM: 603879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC7 PubMed Links

Gene Details

Gene
ARMC7
Gene Name
armadillo repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304271.1 355 Silent Mutation AAA,AAG K6K NP_001291200.1
NM_001304272.1 355 Silent Mutation AAA,AAG K6K NP_001291201.1
NM_001304273.1 355 Silent Mutation AAA,AAG K6K NP_001291202.1
NM_024585.3 355 Silent Mutation AAA,AAG K6K NP_078861.1
Gene
SLC16A5
Gene Name
solute carrier family 16 member 5
There are no transcripts associated with this gene.

View Full Product Details