Product Details

SNP ID
rs145061445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43013142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTTAGTTTCCAATTTGCGGAT[C/G]CACTGCCCTCTGCTTGCGGGCTCTG
Phenotype
MIM: 600735 MIM: 604631
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005533.4 424 Missense Mutation ATC,ATG I72M NP_005524.2
XM_005257302.4 424 Missense Mutation ATC,ATG I72M XP_005257359.1
XM_017024584.1 424 Missense Mutation ATC,ATG I52M XP_016880073.1
XM_017024585.1 424 Missense Mutation ATC,ATG I52M XP_016880074.1
Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details