Product Details

SNP ID
rs145188676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38942842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCCATTCACAGCTTTACTAGCC[A/G]TGAAACTGAAACAGATGTTGTTTCC
Phenotype
MIM: 609498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO47 PubMed Links

Gene Details

Gene
FBXO47
Gene Name
F-box protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008777.2 1292 Missense Mutation ACG,ATG T340M NP_001008777.2
XM_011524865.2 1292 Missense Mutation ACG,ATG T314M XP_011523167.1
XM_011524866.2 1292 Missense Mutation ACG,ATG T283M XP_011523168.1
XM_011524867.2 1292 Missense Mutation ACG,ATG T340M XP_011523169.1

View Full Product Details