Product Details

SNP ID
rs145405642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28573819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATTGAGTGCAGAGGCTTGCAG[C/G]GCACGCCCATAGGAGAAGGTAAGCG
Phenotype
MIM: 103870 MIM: 610271 MIM: 615562
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALDOC PubMed Links

Gene Details

Gene
ALDOC
Gene Name
aldolase, fructose-bisphosphate C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005165.2 1060 Silent Mutation GCC,GCG A305A NP_005156.1
XM_005257949.2 1060 Silent Mutation GCC,GCG A305A XP_005258006.1
XM_011524556.1 1060 Silent Mutation GCC,GCG A305A XP_011522858.1
Gene
PIGS
Gene Name
phosphatidylinositol glycan anchor biosynthesis class S
There are no transcripts associated with this gene.

Gene
SPAG5
Gene Name
sperm associated antigen 5
There are no transcripts associated with this gene.

View Full Product Details