Product Details

SNP ID
rs145526769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7462167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACCGGGAGAGCGGCCAAGTTAC[C/T]GAAGTTGTAAGGGTAGGCGGCAAGG
Phenotype
MIM: 100710 MIM: 612308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB1 PubMed Links

Gene Details

Gene
CHRNB1
Gene Name
cholinergic receptor nicotinic beta 1 subunit
There are no transcripts associated with this gene.

Gene
ZBTB4
Gene Name
zinc finger and BTB domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128833.1 3424 Missense Mutation AGT,GGT S939G NP_001122305.1
NM_020899.3 3424 Missense Mutation AGT,GGT S939G NP_065950.2
XM_006721563.3 3424 Missense Mutation AGT,GGT S939G XP_006721626.1
XM_006721564.2 3424 Missense Mutation AGT,GGT S939G XP_006721627.1
XM_011523972.2 3424 Missense Mutation AGT,GGT S939G XP_011522274.1

View Full Product Details