Product Details

SNP ID
rs146102857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56978231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCCGCTGCTGCTGGGCCTGAA[C/G]GCAGGTAGGTTCAAGCAAGAGGCGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCPEP1 PubMed Links

Gene Details

Gene
SCPEP1
Gene Name
serine carboxypeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021626.2 107 Missense Mutation AAC,AAG N24K NP_067639.1
XM_005257557.2 107 Missense Mutation AAC,AAG N24K XP_005257614.1

View Full Product Details