Product Details
- SNP ID
-
rs146267940
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:3197767 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTAACCATCCCTAAGGTGCTGGC[A/C]AACCATCTCTTGGGGAGCAAGTTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR1A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2241092] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR1A2
- Gene Name
- olfactory receptor family 1 subfamily A member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012352.2 |
251 |
Silent Mutation |
GCA,GCC |
A83A |
NP_036484.1 |
View Full Product Details