Product Details

SNP ID
rs146484834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56834966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCGCAGGGACATCTTCCGCAA[A/G]AGCAAGCACGGGTGGCGCGACACGG
Phenotype
MIM: 601440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085430.2 351 Intron NP_001078899.2
XM_011524735.2 351 Intron XP_011523037.1
Gene
DGKE
Gene Name
diacylglycerol kinase epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003647.2 351 Silent Mutation AAA,AAG K57K NP_003638.1
XM_011525394.2 351 Silent Mutation AAA,AAG K75K XP_011523696.1
XM_011525395.2 351 Silent Mutation AAA,AAG K75K XP_011523697.1
XM_011525396.2 351 Silent Mutation AAA,AAG K75K XP_011523698.1
XM_017025243.1 351 Silent Mutation AAA,AAG K57K XP_016880732.1
XM_017025244.1 351 Silent Mutation AAA,AAG K75K XP_016880733.1
XM_017025245.1 351 Silent Mutation AAA,AAG K75K XP_016880734.1

View Full Product Details