Product Details

SNP ID
rs146574131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8074133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTGCCCCGACTCAACCCAGC[A/G]CCCCCTCTCCACTCCCTCCCATCAC
Phenotype
MIM: 603741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139.2 Intron NP_001130.1

View Full Product Details